missing translation for 'onlineSavingsMsg'
Learn More

Alamethicin, ≥50%, MP Biomedicals™

Product Code. 11484292 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484292 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11484292 Supplier MP Biomedicals Supplier No. 0215900905
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Type Alamethicin
Physical Form Powder
CAS 59588-86-2
Color Tan
Quantity 5 mg
Assay Percent Range ≥50%
Molecular Formula C92H150N22O25
Synonym U-22324 Antibiotic
InChI Key LGHSQOCGTJHDIL-SLKIUSOBSA-N
SMILES CC(C)CC(C(=O)NC(C)(C)C(=O)N1CCCC1C(=O)NC(C(C)C)C(=O)NC(C)(C)C(=O)NC(C)(C)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)N)C(=O)NC(CC2=CC=CC=C2)CO)NC(=O)CNC(=O)C(C)(C)NC(=O)C(C(C)C)NC(=O)C(C)(C)NC(=O)C(CCC(=O)N)NC(=O)C(C)NC(=O)C(C)(C)NC(=O)C(C)NC(=O)C(C)(C)NC(=O)C3CCCN3C
Molecular Weight (g/mol) 1964.341
PubChem CID 53229968
Formula Weight 1964.35
Chemical Name or Material Alamethicin
Show More Show Less
compliance-icons
Product Identifier
  • Alamethicin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contain: Alamethicin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.