missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alas1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
206.00 € - 463.00 €
Specifications
| Antigen | Alas1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18654372
|
Novus Biologicals
NBP2-92476-0.02ml |
0.02 mL |
206.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654962
|
Novus Biologicals
NBP2-92476-0.1ml |
0.1 mL |
463.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alas1 Polyclonal antibody specifically detects Alas1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Alas1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 211 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 5-aminolevulinic acid synthase 1, ALAS, ALAS3, ALASH, ALAS-H, aminolevulinate, delta-, synthase 1, Delta-ALA synthase 1,5-aminolevulinate synthase, nonspecific, mitochondrial, Delta-aminolevulinate synthase 1, EC 2.3.1, EC 2.3.1.37, MIG4, migration-inducing protein 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human ALAS1 (NP_954635.1). MESVVRRCPFLSRVPQAFLQKAGKSLLFYAQNCPKMMEVGAKPAPRALSTAAVHYQQIKETPPASEKDKTAKAKVQQTPDGSQQSPDGTQLPSGHPLPATSQGTASKCPFLAAQMNQRGSSVFCKASLELQEDVQEMNAVRKEVAETSAGPSVVSVKTDGGDPSGLLKNFQDIMQKQRPERVSHLLQDNLPKSVSTFQYDRFFEKKIDEKKNDHTYRVFKTVNRRAHIFPMADDYSDSLITKKQVSVWCS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title