missing translation for 'onlineSavingsMsg'
Learn More

Albendazole, MedChemExpress

Product Code. 16498176
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498176 100 mg 100mg
16462066 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498176 Supplier Medchem Express Supplier No. HYB0223
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Albendazole (SKF-62979) is an orally active and broad-spectrum parasiticide with high effectiveness and low host toxicity, is used for the research of gastrointestinal parasites in humans and animals. Albendazole induces apoptosis and autophagy in cancer cells. Albendazole also inhibits tubulin polymerization and HIF-1α, VEGF expression, has antioxidant activity, and inhibits the glycolytic process in cancer cells.

Albendazole (SKF-62979) is an orally active and broad-spectrum parasiticide with high effectiveness and low host toxicity, is used for the research of gastrointestinal parasites in humans and animals. Albendazole induces apoptosis and autophagy in cancer cells. Albendazole also inhibits tubulin polymerization and HIF-1α, VEGF expression, has antioxidant activity, and inhibits the glycolytic process in cancer cells.

Specifications

Chemical Name or Material Albendazole
Physical Form Solid
CAS 54965-21-8
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H15N3O2S
For Use With (Application) COVID-19-immunoregulation
Synonym SKF-62979
Solubility Information DMSO : 20 mg/mL (75.38 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.53 mM; Need ultrasonic)
SMILES O=C(OC)NC1=NC2=CC=C(SCCC)C=C2N1
Molecular Weight (g/mol) 265.33
Formula Weight 265.33
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.