missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Albumin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
196.00 € - 231.00 €
Specifications
| Antigen | Albumin |
|---|---|
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18247952
|
Novus Biologicals
NB600-570-0.05ML |
0.05 mL |
196.00 €
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18740704
|
Novus Biologicals
NB600-570 |
231.00 €
0.25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Albumin Polyclonal specifically detects Albumin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Albumin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| ALB | |
| IgG | |
| Protein G purified | |
| This reacts with human albumin. This is essentially free of other rabbit serum proteins. Specificity is determined by Ouchterlony Double Diffusion (ODD) and immunoelectrophoresis (IEP) versus human serum and human albumin. |
| Immunohistochemistry (Paraffin), Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers | |
| P02768 | |
| 213 | |
| Purified human albumin. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title