missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ alcohol dehydrogenase 1C Protein

Product Code. p-200042166 Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
100μg
Unit Size:
0.5mg
100µg
This item is not returnable. View return policy

Product Code. 18494391

Brand: Novus Biologicals™ NBP1989360.5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-375 of Human alcohol dehydrogenase 1C The Recombinant Human alcohol dehydrogenase 1C Protein is derived from E. coli. The Recombinant Human alcohol dehydrogenase 1C Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_000660
Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer, pH 8.0, 10% glycerol, 2 mM DTT, 200 mM NaCl
Gene ID (Entrez) 126
Purification Method SDS-PAGE
Quantity 0.5 mg
Source Recombinant human ADH1C protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen ADH1C, 1-375 aa, Human, His tag, E. coli (NP_000660).
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name alcohol dehydrogenase 1 C
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.