missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldehydes Mixture 875 1000 μg/mL in Water, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Acetaldehyde∣Paraformaldehyde |
| CAS | 75-07-0, 30525-89-4 |
| Analyte or Component Names | Acetaldehyde∣Paraformaldehyde |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Water |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Acetaldehyde 1000 μg/mL∣Paraformaldehyde 1000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Individuate un'opportunità di miglioramento?