missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH1A1 Mouse, Clone: ALDH1A1/1381, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 216-MSM1-P1
This item is not returnable.
View return policy
Specifications
| ALDH1A1 | |
| Cytoplasmic | |
| ALDH1A1/1381 | |
| Unconjugated | |
| NP_000680 | |
| Mouse | |
| 56kDa | |
| Protein A/G Purified | |
| RUO | |
| 216 | |
| IgG1 |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| ALDH1A1 | |
| Purified recombinant fragment of human ALDH1A1 (aa 315-434) | |
| HepG2 or A549 cells. Colon or Testicular Carcinoma. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction