Learn More
ALDH1A3 Antibody (CL11968) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-15126JF646
Product Code. 30301908
  • 737.00 € / 0.10mL
Description
ALDH1A3 Monoclonal antibody specifically detects ALDH1A3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ALDH1A3
Monoclonal
Janelia Fluor 646
aldehyde dehydrogenase 1 family, member A3, Aldehyde dehydrogenase 6, aldehyde dehydrogenase family 1 member A3, ALDH1A6, ALDH6acetaldehyde dehydrogenase 6, EC 1.2.1, EC 1.2.1.5, RalDH3, RALDH-3, Retinaldehyde dehydrogenase 3
This antibody was developed using a synthetic peptide derived from P47895, with the exact immunogen sequence remaining proprietary.
0.1 mL
Vision
220
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL11968
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified