missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH1L2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92166-0.1ml
This item is not returnable.
View return policy
Description
ALDH1L2 Polyclonal antibody specifically detects ALDH1L2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ALDH1L2 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000 | |
| aldehyde dehydrogenase 1 family, member L2, aldehyde dehydrogenase family 1 member L2, DKFZp686A16126, DKFZp686M064, EC 1.5.1.6, FLJ36769, FLJ38508, MGC119536, MGC119537,10-formyltetrahydrofolate dehydrogenase ALDH1L2, Mitochondrial 10-formyltetrahydrofolate dehydrogenase, mtFDHDKFZp686P14145, probable 10-formyltetrahydrofolate dehydrogenase ALDH1L2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 280-480 of human ALDH1L2 (NP_001029345.2). EPLEIKGAKKPGLVTKNGLVLFGNDGKALTVRNLQFEDGKMIPASQYFSTGETSVVELTAEEVKVAETIKVIWAGILSNVPIIEDSTDFFKSGASSMDVARLVEEIRQKCGGLQLQNEDVYMATKFEGFIQKVVRKLRGEDQEVELVVDYISKEVNEIMVKMPYQCFINGQFTDADDGKTYDTINPTDGSTICKVSYASLA | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 160428 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction