missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALDH4A1 Recombinant Mouse Monoclonal Antibody (7E2-R)

Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556491
This item is not returnable.
View return policy
Description
ALDH4A1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
ALDH4A1 belongs to the aldehyde dehydrogenase family of proteins. This enzyme is a mitochondrial matrix NAD-dependent dehydrogenase which catalyzes the second step of the proline degradation pathway, converting pyrroline-5-carboxylate to glutamate. Deficiency of this enzyme is associated with type II hyperprolinemia, an autosomal recessive disorder characterized by accumulation of delta-1-pyrroline-5-carboxylate (P5C) and proline.
Specifications
| ALDH4A1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P0C2X9, P30038, Q8CHT0 | |
| Aldh4a1 | |
| Synthetic peptide within Human ALDH4A1 aa 514-563/563. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 7E2-R | |
| Unconjugated | |
| Aldh4a1 | |
| A930035F14Rik; Ahd1; Ahd-1; aldehyde dehydrogenase 4 family member A1; aldehyde dehydrogenase 4 family, member A1; aldehyde dehydrogenase 4A1; aldehyde dehydrogenase family 4 member A1; ALDH4; aldh4a1; Aldh5a1; delta-1-pyrroline-5-carboxylate dehydrogenase, mitochondrial; E330022C09; L-glutamate gamma-semialdehyde dehydrogenase; mitochondrial delta-1-pyrroline 5-carboxylate dehydrogenase; P5C dehydrogenase; P5cd; P5CDh; P5cdhl; P5cdhs; RP11-128M10.1; similar to aldehyde dehydrogenase 4 family, member A1; Ssdh1; zgc:63592 | |
| Mouse | |
| Protein A | |
| RUO | |
| 212647, 641316, 8659 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering