missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldo-keto Reductase 1C3/AKR1C3 Rabbit anti-Human, Mouse, Clone: 7T10F5, Novus Biologicals™
Rabbit Monoclonal Antibody
201.00 € - 510.00 €
Specifications
| Antigen | Aldo-keto Reductase 1C3/AKR1C3 |
|---|---|
| Clone | 7T10F5 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18315305
|
Novus Biologicals
NBP3-16288-20UL |
20 μg |
201.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18311684
|
Novus Biologicals
NBP3-16288-100UL |
100 μg |
510.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aldo-keto Reductase 1C3/AKR1C3 Monoclonal antibody specifically detects Aldo-keto Reductase 1C3/AKR1C3 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| Aldo-keto Reductase 1C3/AKR1C3 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 8644 | |
| IgG | |
| Affinity purified |
| 7T10F5 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 17-beta-hydroxysteroid dehydrogenase type 5, 3-alpha-HSD type 2, 3-alpha-hydroxysteroid dehydrogenase type 2, aldo-keto reductase family 1 member C3, aldo-keto reductase family 1, member C3 (3-alpha hydroxysteroid dehydrogenase, type II), Chlordecone reductase homolog HAKRb, DD-3, DD317-beta-HSD 5, DDH1, DDX, Dihydrodiol dehydrogenase 3, Dihydrodiol dehydrogenase type I, dihydrodiol dehydrogenase X, EC 1.1.1, EC 1.1.1.112,3-alpha-HSD type II, brain, EC 1.1.1.188, EC 1.1.1.213, EC 1.1.1.63, EC 1.1.1.64, EC 1.3.1.20, HA1753, HAKRBtype IIb 3-alpha hydroxysteroid dehydrogenase, HAKRe, HSD17B5, hydroxysteroid (17-beta) dehydrogenase 5, Indanol dehydrogenase, KIAA0119hluPGFS, PGFS, Prostaglandin F synthase, Testosterone 17-beta-dehydrogenase 5, Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human Aldo-keto Reductase 1C3/AKR1C3 (P42330). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title