missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldolase C Antibody (E9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50056
This item is not returnable.
View return policy
Description
Aldolase C Monoclonal antibody specifically detects Aldolase C in Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Aldolase C | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:10000, Immunocytochemistry/ Immunofluorescence 1:1000 - 1:5000 | |
| P09972 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
| Western Blot, Immunofluorescence | |
| E9 | |
| Unconjugated | |
| 50% PBS, 50% glycerol | |
| ALDC, aldolase 3, aldolase C, fructose-bisphosphate, Brain-type aldolase, EC 4.1.2.13, fructoaldolase C, fructose-1,6-biphosphate triosephosphate lyase, fructose-bisphosphate aldolase C | |
| Full length recombinant human Aldolase C. [UniProt# P09972] | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 230 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction