missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldolase C Mouse anti-Human, Mouse, Clone: 2.00E+11, Novus Biologicals™
Mouse Monoclonal Antibody
304.00 € - 486.00 €
Specifications
| Antigen | Aldolase C |
|---|---|
| Clone | 200000000000 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1018, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18237221
|
Novus Biologicals
NBP2-59408-50UL |
50 μL |
304.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18204341
|
Novus Biologicals
NBP2-59408-100UL |
100 μL |
486.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aldolase C Monoclonal specifically detects Aldolase C in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Aldolase C | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Core ESC Like Genes, Stem Cell Markers | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 230 | |
| Recombinant human ALDOC (1-364aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 200000000000 | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1018, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ALDC, aldolase 3, aldolase C, fructose-bisphosphate, Brain-type aldolase, EC 4.1.2.13, fructoaldolase C, fructose-1,6-biphosphate triosephosphate lyase, fructose-bisphosphate aldolase C | |
| ALDOC | |
| IgG1 κ | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title