missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALG9 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92351-0.1ml
This item is not returnable.
View return policy
Description
ALG9 Polyclonal antibody specifically detects ALG9 in Human samples. It is validated for Western Blot
Specifications
| ALG9 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| alpha-1,2-mannosyltransferase ALG9, asparagine-linked glycosylation 9 homolog (S. cerevisiae, alpha-1,2-mannosyltransferase), asparagine-linked glycosylation 9 homolog (yeast, alpha 1,2mannosyltransferase), asparagine-linked glycosylation 9 homolog (yeast, alpha-1,2-mannosyltransferase), asparagine-linked glycosylation 9, alpha- 1,2-mannosyltransferase homolog (S.cerevisiae, alpha- 1,2-mannosyltransferase), asparagine-linked glycosylation 9, alpha-1,2-mannosyltransferase homolog (S.cerevisiae), Asparagine-linked glycosylation protein 9 homolog, CDG1L, DIBD1FLJ21845, disrupted in bipolar affective disorder 1, Disrupted in bipolar disorder protein 1, DKFZp586M2420, EC 2.4.1.-, LOH11CR1J, loss of heterozygosity, 11, chromosomal region 1 gene J product | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 439-618 of human ALG9 (NP_079016.2). LFRGYHGPLDLYPEFYRIATDPTIHTVPEGRPVNVCVGKEWYRFPSSFLLPDNWQLQFIPSEFRGQLPKPFAEGPLATRIVPTDMNDQNLEEPSRYIDISKCHYLVDLDTMRETPREPKYSSNKEEWISLAYRPFLDASRSSKLLRAFYVPFLSDQYTVYVNYTILKPRKAKQIRKKSGG | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79796 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction