missing translation for 'onlineSavingsMsg'
Learn More

Alisertib, MedChemExpress

Product Code. 16497037
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497037 5 mg 5mg
16495236 100 mg 100mg
16471006 200 mg 200mg
16470647 50 mg 50mg
16414328 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497037 Supplier Medchem Express Supplier No. HY10971
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Alisertib (MLN 8237) is an orally active and selective Aurora A kinase inhibitor (IC50=1.2 nM), which binds to Aurora A kinase resulting in mitotic spindle abnormalities, mitotic accumulation. Alisertib (MLN 8237) induces apoptosis and autophagy through targeting the AKT/mTOR/AMPK/p38 pathway in leukemic cells. Antitumor activity.

Alisertib (MLN 8237) is an orally active and selective Aurora A kinase inhibitor (IC50=1.2 nM), which binds to Aurora A kinase resulting in mitotic spindle abnormalities, mitotic accumulation. Alisertib (MLN 8237) induces apoptosis and autophagy through targeting the AKT/mTOR/AMPK/p38 pathway in leukemic cells. Antitumor activity.

Tekniske data

Chemical Name or Material Alisertib
CAS 1028486-01-2
Color Light Nude
Purity Grade Notes Research
Molecular Formula C27H20ClFN4O4
Quantity 5 mg
Synonym MLN 8237
Solubility Information DMSO : 25 mg/mL (48.18 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C=C1OC)NC2=NC=C3CN=C(C4=CC(Cl)=CC=C4C3=N2)C5=C(C=CC=C5F)OC)O
Molecular Weight (g/mol) 518.92
Formula Weight 518.92
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.