missing translation for 'onlineSavingsMsg'
Learn More

Aliskiren, MedChemExpress

Product Code. 16451247
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451247 50 mg 50mg
16435008 10 mg 10mg
16427737 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16451247 Supplier Medchem Express Supplier No. HY12176
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Aliskiren is an orally active, highly potent and selective renin inhibitor, with IC50 of 1.5 nM. Aliskiren can be used for the research of hypertension, cardiovascular diseases and cancer cachexia.

Aliskiren is an orally active, highly potent and selective renin inhibitor, with IC50 of 1.5 nM. Aliskiren can be used for the research of hypertension, cardiovascular diseases and cancer cachexia.

Specifications

Chemical Name or Material Aliskiren
CAS 173334-57-1
Color Colorless
Purity Grade Notes Research
Molecular Formula C30H53N3O6
Quantity 50 mg
Synonym CGP 60536 CGP60536B SPP 100
Solubility Information Ethanol : 100 mg/mL (181.24 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (181.24 mM; Need ultrasonic)
SMILES COCCCOC1=CC(C[C@@H](C[C@@H]([C@H](C[C@H](C(NCC(C)(C(N)=O)C)=O)C(C)C)O)N)C(C)C)=CC=C1OC
Molecular Weight (g/mol) 551.76
Formula Weight 551.76
Percent Purity 95.24%
Grade Research
Physical Form Oil
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.