Learn More
Novus Biologicals™ ALK-1 Antibody (ascrinvacumab) - IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28297-1mg
 View more versions of this product
Product Code. 30047517
  • 920.00 € / 1mg
Description
ALK-1 Monoclonal antibody specifically detects ALK-1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ALK-1
Monoclonal
Unconjugated
P37023
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG2
Flow Cytometry, ELISA, Functional Assay
ascrinvacumab
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRLK1ORW2, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I
ACVRL1/ ALK-1
1 mg
Phospho Specific, Protein Kinase, Signal Transduction
94
Human
Purified