missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Preparation Method | This product is intended for use as a positive control in Western Blot. Each vial of cell lysate contains 100 μg of total protein which should be sufficient for 20-50 reactions. Depending on over-expression level, antibody affinity and detection system, some lysates can go as low as 0.1 μg per load. We recommend starting with 5 μg of cell lysate. Add an equal amount of cell lysate and 2X SDS Sample buffer and boil the SDS samples for 10 minutes before loading. |
| Quantity | 0.1 mg |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| Research Category | Protein Kinase, Signal Transduction |
| Target | ALK/CD246 |
| Synonym | ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3 |
| Gene Symbol | ALK |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?