missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase, Intestinal Antibody (AP-7/6/7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
201.00 € - 351.00 €
Specifications
| Antigen | Alkaline Phosphatase, Intestinal |
|---|---|
| Clone | AP-7/6/7 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence |
| Applications | Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18616126
|
Novus Biologicals
NBP2-50102-0.025ml |
0.025 mL |
201.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651148
|
Novus Biologicals
NBP2-50102 |
0.1 mL |
351.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alkaline Phosphatase, Intestinal Monoclonal antibody specifically detects Alkaline Phosphatase, Intestinal in Bovine samples. It is validated for Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Alkaline Phosphatase, Intestinal | |
| 1 mg/mL | |
| Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Ovarian Carcinoma Cell Markers, Stem Cell Markers | |
| PBS | |
| 248 | |
| IgG1 | |
| Protein A purified |
| AP-7/6/7 | |
| Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Bovine | |
| alkaline phosphatase, intestinal, alkaline phosphomonoesterase, EC 3.1.3.1, glycerophosphatase, IAP, Intestinal alkaline phosphatase, intestinal-type alkaline phosphatase, Kasahara isozyme | |
| Calf intestinal alkaline phosphatase | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title