missing translation for 'onlineSavingsMsg'
Learn More

Alkaline Phosphatase Mouse, Clone: ALPL/597, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 249-MSM1-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960048

  • 485.00 € / 100µg
Estimated Shipment: 03-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Alkaline Phosphatase
Cell Surface
ALPL/597
Unconjugated
NP_016856392
Mouse
55kDa
Protein A/G Purified
RUO
249
IgG1 κ
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
ALPL
Recombinant human ALPL protein
Intestine
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers