missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALKBH8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 550.00 €
Specifications
| Antigen | ALKBH8 |
|---|---|
| Dilution | Western Blot 1:2000 - 1:10000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233125
|
Novus Biologicals
NBP3-38393-20ul |
20 μL |
190.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231493
|
Novus Biologicals
NBP3-38393-100ul |
100 μL |
550.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALKBH8 Polyclonal antibody specifically detects ALKBH8 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| ALKBH8 | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| ABH8MGC10235, AlkB homologue 8, alkB, alkylation repair homolog 8 (E. coli), alkylated DNA repair protein alkB homolog 8, EC 1.14.11.-, EC 2.1.1.-, FLJ38204, Probable alpha-ketoglutarate-dependent dioxygenase ABH8, S-adenosyl-L-methionine-dependent tRNA methyltransferase ABH8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human ALKBH8 (NP_620130.2).,, Sequence:, MDSNHQSNYKLSKTEKKFLRKQIKAKHTLLRHEGIETVSYATQSLVVANGGLGNGVSRNQLLPVLEKCGLVDALLMPPNKPYSFARYRTTEESKRAYVTLNGKEVVDDLGQKITLYLNFVEKVQWKELRPQALPPGLMVVEEIISSEEEKMLLESVDWTEDTDNQNSQKSLKHRRVKHFGYEFHYENNNVDKDKPLSGGLPDICESFLEKWLRKGYIKHKPDQMTINQYE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:2000 - 1:10000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 91801 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title