missing translation for 'onlineSavingsMsg'
Learn More

Grace™ Alltech™ Vydac™ 214TP C4 Column

Product Code. 11349640
Encompass
Click to view available options
Length (Metric):
150 mm
Diameter (Metric):
4.6 mm
Particle Size:
5 μm
This item is not returnable. View return policy

Product Code. 11349640

Brand: Grace™ 214TP5415

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

A polymerically bonded endcapped n-butyl reversed-phase based on 300Å TP silica

  • Polymeric bonded phases have exceptionally long column lifetime and negligible phase leaching
  • Extensive applications library based on over two decade's experience

Specifications

Diameter (Metric) 4.6 mm
For Use With Glycoproteins, Hemoglobin Variants, Histones, Human Growth Hormone, Insulin Variants, Membrane Proteins, Analysis and Purification of Proteins and Peptides
Length (Metric) 150 mm
Particle Shape Spherical
Particle Size 5 μm
Pore Size 300 Å
Surface Area 60 to 110 m2/g
USP Type L26
Stationary Phase 214 TP
Packing Material Silica
Carbon Load 3%
Column Type C4
Endcapped Yes
Type Column
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.