missing translation for 'onlineSavingsMsg'
Learn More

Allyl Bromide (Stabilized with Propylene Oxide), TRC

Product Code. 30134028
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134028 10 g 10g
30133912 50 g 50g
30011802 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134028 Supplier Toronto Research Chemicals Supplier No. TRCA54965510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-95-6
Molecular Formula C3 H5 Br
Molecular Weight (g/mol) 120.98
Synonym 3-Bromo-1-propene, 1-Bromo-2-propene, 2-Propenyl Bromide, 3-Bromo-1-propene, 3-Bromopropene, 3-Bromopropylene, Allyl Bromide, NSC 7596
IUPAC Name 3-bromoprop-1-ene
SMILES BrCC=C

Specifications

Analyte or Component Names Allyl Bromide
Shipping Condition Blue Ice
InChI Formula InChI=1S/C3H5Br/c1-2-3-4/h2H,1,3H2
Solution Type Neat
Industry Type Pharmaceutical
Quantity 10 g
Formula Weight 119.9575
Percent Purity >95%
Chemical Name or Material Allyl Bromide
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.