missing translation for 'onlineSavingsMsg'
Learn More

Allyl phenyl ether, 99%

Product Code. 11312116
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312116 25 g 25g
11322116 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312116 Supplier Thermo Scientific Alfa Aesar Supplier No. L03359.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1746-13-0 | C9H10O | 134.178 g/mol

Allyl phenyl ether is an pharmaceutical and OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl phenyl ether is an pharmaceutical and OLED intermediate.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents, acids, bases. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1746-13-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00008644
InChI Key POSICDHOUBKJKP-UHFFFAOYSA-N
Synonym allyl phenyl ether, allyloxy benzene, allyloxybenzene, benzene, 2-propenyloxy, allyl phenoxylate, phenyl allyl ether, ether, allyl phenyl, 3-phenoxypropene, phenylpropenyl ether, phenyl 2-propenyl ether
PubChem CID 74458
IUPAC Name prop-2-enoxybenzene
SMILES C=CCOC1=CC=CC=C1

Specifications

Density 0.979
Boiling Point 191°C to 192°C
Flash Point 62°C (143°F)
Odor Characteristic
Refractive Index 1.521
Quantity 25 g
Beilstein 1905622
Solubility Information Insoluble in water.
Formula Weight 134.18
Percent Purity 99%
Chemical Name or Material Allyl phenyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.