missing translation for 'onlineSavingsMsg'
Learn More

Allylmagnesium chloride, 1.7M solution in THF, AcroSeal™

Product Code. 10372373 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10372373 100 mL AcroSeal™ Glass bottle 100mL
10464643 800 mL AcroSeal™ Glass bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10372373 Supplier Thermo Scientific Acros Supplier No. 209671000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 11-05-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 2622-05-1 | C3H5ClMg | 100.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2622-05-1 , 592-42-7
Molecular Formula C3H5ClMg
Molecular Weight (g/mol) 100.83
MDL Number MFCD00000473
InChI Key PLYLAFITHJUEGX-UHFFFAOYSA-M
SMILES Cl[Mg]CC=C

Specifications

Color Black
CAS Min % 15.76, 80.04
CAS Max % 19.96, 84.25
Concentration 1.5 to 1.9M
Density 0.9600g/mL
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.96
Linear Formula H2C=CHCH2MgCl
Quantity 100 mL
Flash Point −17°C
Physical Form Liquid
Synonym tri-o-tolyl phosphite, tri-o-tolylphosphite, tri-o-cresyl phosphite, phosphorous acid tri-o-cresyl ester, tris o-methylphenyl phosphite, tris 2-methylphenyl phosphite, tris 2-tolyl phosphite, tris o-tolyloxy phosphine, phosphorous acid, tri-o-cresyl ester, tri-2-tolylphosphite
Solubility Information Solubility in water: vigorous reaction.
IUPAC Name tris(2-methylphenyl) phosphite
PubChem CID 92937
Formula Weight 100.83
Percent Purity 1.5 to 1.9M
Chemical Name or Material Allylmagnesium chloride, 1.7M Solution in THF
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allylmagnesium chloride, 1.7M solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.