missing translation for 'onlineSavingsMsg'
Learn More

Allylpentafluorobenzene, 97%, Thermo Scientific™

Product Code. 15412142 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15412142 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15412142 Supplier Thermo Scientific Acros Supplier No. 310850250
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 1736-60-3
Melting Point -64.0°C
Density 1.3580g/mL
Boiling Point 148.0°C to 149.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Molecular Formula C9H5F5
Linear Formula C6F5CH2CH=CH2
MDL Number MFCD00000301
Quantity 25 g
Specific Gravity 1.358
Synonym allylpentafluorobenzene, benzene, allylpentafluoro, 3-pentafluorophenyl-1-propene, 2,3,4,5,6-pentafluoro allylbenzene, benzene, pentafluoro-2-propenyl, 1,2,3,4,5-pentafluoro-6-prop-2-en-1-yl benzene, allyl pentafluorobenzene, pubchem13366, acmc-1bth5
InChI Key YBDBTBVNQQBHGJ-UHFFFAOYSA-N
SMILES FC1=C(F)C(F)=C(CC=C)C(F)=C1F
IUPAC Name 1,2,3,4,5-pentafluoro-6-(prop-2-en-1-yl)benzene
Molecular Weight (g/mol) 208.13
PubChem CID 74435
Formula Weight 208.13
Percent Purity 97%
Chemical Name or Material Allylpentafluorobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.