missing translation for 'onlineSavingsMsg'
Learn More

Allylpentafluorobenzene, 97%, Thermo Scientific™

Product Code. 10421691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10421691 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10421691 Supplier Thermo Scientific Acros Supplier No. 310850050
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 1736-60-3
CAS Min % 96.0
CAS Max % 100.0
Melting Point -64.0°C
Color Colorless
Density 1.3580g/mL
Boiling Point 148.0°C to 149.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C9H5F5
Linear Formula C6F5CH2CH=CH2
Refractive Index 1.4270 to 1.4290 (20°C, 589nm)
MDL Number MFCD00000301
Quantity 5 g
Specific Gravity 1.358
Synonym allylpentafluorobenzene, benzene, allylpentafluoro, 3-pentafluorophenyl-1-propene, 2,3,4,5,6-pentafluoro allylbenzene, benzene, pentafluoro-2-propenyl, 1,2,3,4,5-pentafluoro-6-prop-2-en-1-yl benzene, allyl pentafluorobenzene, pubchem13366, acmc-1bth5
InChI Key YBDBTBVNQQBHGJ-UHFFFAOYSA-N
SMILES FC1=C(F)C(F)=C(CC=C)C(F)=C1F
IUPAC Name 1,2,3,4,5-pentafluoro-6-prop-2-enylbenzene
Molecular Weight (g/mol) 208.13
PubChem CID 74435
Formula Weight 208.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Allylpentafluorobenzene, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.