missing translation for 'onlineSavingsMsg'
Learn More

Allyltriphenylphosphonium bromide, 99%

Product Code. 10655132 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10655132 100 g Glass bottle 100g
10456422 5 g Glass bottle 5g
10316310 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10655132 Supplier Thermo Scientific Acros Supplier No. 103041000
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1560-54-9 | C21H20BrP | 383.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1560-54-9
Molecular Formula C21H20BrP
Molecular Weight (g/mol) 383.27
MDL Number MFCD00011808
InChI Key FWYKRJUVEOBFGH-UHFFFAOYSA-M
Synonym allyltriphenylphosphonium bromide, triphenyl allylphosphonium bromide, phosphonium, triphenyl-2-propenyl-, bromide, allyl triphenylphosphonium bromide, phosphonium, allyltriphenyl-, bromide, allyltriphenylphosphonium bromide tal, triphenyl prop-2-enyl phosphanium bromide, 2-propenyltriphenylphosphonium bromide, triphenyl prop-2-en-1-yl phosphanium bromide, wln: 1u2pr&r&r &e
PubChem CID 197740
IUPAC Name triphenyl(prop-2-enyl)phosphanium;bromide
SMILES [Br-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 222.0°C to 225.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2C=CHCH2P(C6H5)3Br
Quantity 100 g
Solubility Information Solubility in water: decomposes
Formula Weight 383.26
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Allyltriphenylphosphonium bromide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Allyltriphenylphosphonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.