missing translation for 'onlineSavingsMsg'
Learn More

Alodane, TRC

Product Code. 30138187
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138187 50 mg 50mg
30133451 100 mg 100mg
30004495 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138187 Supplier Toronto Research Chemicals Supplier No. TRCA61428350MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2550-75-6
Molecular Formula C9 H6 Cl8
Molecular Weight (g/mol) 397.77
Synonym 2-Norbornene, 1,2,3,4,7,7-hexachloro-5,6-bis(chloromethyl)- (6CI,7CI,8CI), 1,2,3,4,7,7-Hexachloro-5,6-bis(chloromethyl)bicyclo[2.2.1]hept-2-ene, 1,2,3,4,7,7-Hexachloro-5,6-bis(chloromethyl)-2-norbornene, 1,2,3,4,7,7-Hexachloro-5,6-di(chloromethyl)[2.2.1]bicyclohept-2-ene, AC 12402, Alodan, Alodan (pesticide), Chlorbicyclen, Cyclodan, Hercules 12402, Hercules 426, NSC 41891
IUPAC Name (1R,4S)-1,2,3,4,7,7-hexachloro-5,6-bis(chloromethyl)bicyclo[2.2.1]hept-2-ene
SMILES ClCC1C(CCl)[C@]2(Cl)C(=C(Cl)[C@@]1(Cl)C2(Cl)Cl)Cl

Specifications

Analyte or Component Names Chlorbicyclen
Shipping Condition Room Temperature
InChI Formula InChI=1S/C9H6Cl8/c10-1-3-4(2-11)8(15)6(13)5(12)7(3,14)9(8,16)17/h3-4H,1-2H2/t3?,4?,7-,8+
Solution Type Neat
Industry Type Pharmaceutical
Quantity 50 mg
Formula Weight 393.7978
Chemical Name or Material Chlorbicyclen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.