missing translation for 'onlineSavingsMsg'
Learn More

Aloin, MedChemExpress

Product Code. 16494757
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494757 50 mg 50mg
16441071 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16494757 Supplier Medchem Express Supplier No. HYN0123
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Aloin(Aloin-A; Barbaloin-A) is a natural antitumor anthraquinone glycoside with iron chelating and non-atherogenic activities.

Aloin(Aloin-A

Specifications

Chemical Name or Material Aloin
CAS 1415-73-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H22O9
Quantity 50 mg
Synonym Aloin-A Barbaloin-A
Solubility Information DMSO : 125 mg/mL (298.76 mM; Need ultrasonic)
SMILES O=C1C2=C(C=CC=C2O)[C@H]([C@]3([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)C4=CC(CO)=CC(O)=C14
Molecular Weight (g/mol) 418.39
Formula Weight 418.39
Percent Purity 98.21%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.