missing translation for 'onlineSavingsMsg'
Learn More

Aloin

Product Code. 15424579
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424579 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15424579 Supplier Thermo Scientific Alfa Aesar Supplier No. J62153.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Aloe exudant, used as a stimulant-laxative | CAS: 1415-73-2 | C21H22O9 | 418.40 g/mol

Aloe exudant, used as a stimulant-laxative

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1415-73-2
Molecular Formula C21H22O9
Molecular Weight (g/mol) 418.40
MDL Number MFCD00151160
InChI Key AFHJQYHRLPMKHU-WEZNYRQKSA-N
Synonym barbaloin, aloin, aloin a, aloinum, aloin ban, dsstox_cid_25967, 10s-1,8-dihydroxy-3-hydroxymethyl-10-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl-10h-anthracen-9-one, 1s-1,5-anhydro-1-9s-4,5-dihydroxy-2-hydroxymethyl-10-oxo-9,10-dihydroanthracen-9-yl-d-glucitol, 10-1',5'-anhydroglucosyl aloe-emodin-9-anthrone, barbaloin a
PubChem CID 12305761
ChEBI CHEBI:2991
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)[C@@H]1C2=CC=CC(O)=C2C(=O)C2=C(O)C=C(CO)C=C12

Specifications

Color Yellow to Brown
Quantity 25 g
Beilstein 6077558
IUPAC Name (10S)-1,8-dihydroxy-3-(hydroxymethyl)-10-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-10H-anthracen-9-one
Formula Weight 418.4
Physical Form Powder
Chemical Name or Material Aloin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.