Learn More
Invitrogen™ Alomfilimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559084
 View more versions of this product
Product Code. 30283171
  • 1204.00 € / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Alomfilimab is a monoclonal antibody that targets and inhibits interleukin-1 receptor type 1 (IL-1R1), thereby blocking the activity of interleukin-1 (IL-1) and reducing inflammation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Alomfilimab
Recombinant Monoclonal
Unconjugated
KY-1044; SAR-445256
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized