missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ alpha 1-Microglobulin Native Protein

Product Code. 18613287 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18613287 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18613287 Supplier R&D Systems™ Supplier No. NBP262396
  • ONLINE EXCLUSIVE PRICE
    778.58 €
    663.60 € / 1mg
    Save 114.98 €
    14% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Native Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA
Formulation Ammonium bicarbonate
Gene ID (Entrez) 259
Molecular Weight (g/mol) MolecularWeight-theroretical: 27 kDa
Quantity 1 mg
Storage Requirements Store at 4°C. After reconstitution store at -20°C.
Gene Alias A1M, alpha-1-microglobulin/bikunin precursor, bikunin, EDC1, HCPcomplex-forming glycoprotein heterogeneous in charge, HI30, IATIL, inter-alpha-trypsin inhibitor light chain, ITI, ITILC, ITILgrowth-inhibiting protein 19, protein AMBP, protein HC, trypstatin, uristatin, uronic-acid-rich protein, UTI
Gene Symbol AMBP
Reconstitution Reconstitute with 1.0 mL sterile PBS. Care should be taken during reconstitution as the protein may appear as a film (at the bottom of the vial. Gently mix vial after reconstitution.)
Research Category Cancer
Product Line Novus
Purity or Quality Grade >95%, by SDS-PAGE
Protein alpha 1-Microglobulin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.