missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ alpha-1 Microglobulin Polyclonal Antibody
GREENER_CHOICE

Product Code. 13421507
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13421507 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13421507 Supplier Invitrogen™ Supplier No. PA527314

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, human plasma, HepG2 conditioned medium. Predicted reactivity: Cat (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a complex glycoprotein secreted in plasma. The precursor is proteolytically processed into distinct functioning proteins: alpha-1-microglobulin, which belongs to the superfamily of lipocalin transport proteins and may play a role in the regulation of inflammatory processes, and bikunin, which is a urinary trypsin inhibitor belonging to the superfamily of Kunitz-type protease inhibitors and plays an important role in many physiological and pathological processes. This gene is located on chromosome 9 in a cluster of lipocalin genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-1 Microglobulin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene AMBP
Gene Accession No. P02760
Gene Alias A1M; AI194774; alpha 1 microglobulin/bikunin; alpha-1 microglobulin; Alpha-1 microglobulin/bikunin; Alpha-1 microglycoprotein; Alpha-1-microglobulin; alpha-1-microglobulin/bikunin; alpha1-microglobulin/bikunin; alpha-1-microglobulin/bikunin precursor; alpha-1-microglobulin-bikunin; alpha1-microglobulin-bikunin precursor; Ambp; AMBP protein; ASPI; BI-14; Bikunin; complex-forming glycoprotein heterogeneous in charge; Cumulus extracellular matrix-stabilizing factor; EDC1; ESF; growth-inhibiting protein 19; HCP; HI30; HI-30; IATIL; inter-alpha-trypsin inhibitor (protein HC), light; inter-alpha-trypsin inhibitor light chain; Intin4; ITI; ITIL; ITILC; ITI-LC; polyprotein; protein AMBP; Protein HC; Protein Protein HC; trypstatin; ulinastatin; unnamed protein product; Urinary Trypsin Inhibitor; uristatin; uronic-acid-rich protein; UTI
Gene Symbols AMBP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Alpha 1 Microglobulin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.