missing translation for 'onlineSavingsMsg'
Learn More

Alpha-(2,3-Dimethylphenyl)-1-(trityl)-1H-imidazole-4-methanol, TRC

Product Code. 30375885
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30375885 250 mg
30372601 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30375885 Supplier Toronto Research Chemicals Supplier No. TRCD477340250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 176721-01-0
Molecular Formula C31H28N2O
Molecular Weight (g/mol) 444.58
Synonym Medetomidine Impurity 36
SMILES Cc1cccc(C(O)c2cncn2C(c3ccccc3)(c4ccccc4)c5ccccc5)c1C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C31H28N2O/c1-23-13-12-20-28(24(23)2)30(34)29-21-32-22-33(29)31(25-14-6-3-7-15-25,26-16-8-4-9-17-26)27-18-10-5-11-19-27/h3-22,30,34H,1-2H3
Formula Weight 444.2202
Chemical Name or Material alpha-(2,3-Dimethylphenyl)-1-(trityl)-1H-imidazole-4-methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.