missing translation for 'onlineSavingsMsg'
Learn More

alpha,3-Dimethylstyrene

Product Code. 11434710
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434710 250 mg 250mg
11444710 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434710 Supplier Thermo Scientific Alfa Aesar Supplier No. H50760.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1124-20-5 | C10H12 | 132.206 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1124-20-5
Molecular Formula C10H12
Molecular Weight (g/mol) 132.206
MDL Number MFCD08060972
InChI Key XXTQHVKTTBLFRI-UHFFFAOYSA-N
Synonym m,alpha-dimethylstyrene, alpha,3-dimethylstyrene, m-isopropenyltoluene, unii-3ey8gvm0dx, benzene, 1-methyl-3-1-methylethenyl, 3ey8gvm0dx, 1-methyl-3-prop-1-en-2-yl benzene, 1-isopropenyl-3-methyl-benzene, 1-methyl-3-1-methylethenyl benzene, 1-methyl-3-ethenyl benzene
PubChem CID 70759
IUPAC Name 1-methyl-3-prop-1-en-2-ylbenzene
SMILES CC1=CC=CC(=C1)C(=C)C

Specifikationer

Odor Characteristic
Quantity 250 mg
UN Number UN3295
Formula Weight 132.21
Chemical Name or Material alpha,3-Dimethylstyrene

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.