missing translation for 'onlineSavingsMsg'
Learn More

alpha,4-Dimethylstyrene, stab. with 4-tert-butylcatechol

Product Code. 15407885
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15407885 25 g 25g
15497875 5 g 5g
15417885 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15407885 Supplier Thermo Scientific Alfa Aesar Supplier No. H31739.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1195-32-0 | C10H12 | 132.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1195-32-0
Molecular Formula C10H12
Molecular Weight (g/mol) 132.21
MDL Number MFCD00036510
InChI Key MMSLOZQEMPDGPI-UHFFFAOYSA-N
Synonym 1-methyl-4-prop-1-en-2-yl benzene, dehydro-p-cymene, 2-p-tolylpropene, 4-isopropenyltoluene, 4-methylisopropenylbenzene, 2-p-methylphenyl propene, p-cymenene, p,alpha-dimethylstyrene, p-alpha-dimethylstyrene, 1-isopropenyl-4-methylbenzene
PubChem CID 62385
SMILES CC(=C)C1=CC=C(C)C=C1

Specifications

Density 0.872
Boiling Point 186°C to 189°C
Flash Point 63°C (145°F)
Refractive Index 1.509
Quantity 25 g
IUPAC Name 1-methyl-4-(prop-1-en-2-yl)benzene
Formula Weight 132.21
Chemical Name or Material alpha,4-Dimethylstyrene, stabilized with 4-tert-butylcatechol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.