missing translation for 'onlineSavingsMsg'
Learn More

Alpha-(4-Isobutylphenyl)ethanol, TRC

Product Code. 17912655
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17912655 100 mg 100mg
17902085 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17912655 Supplier Toronto Research Chemicals Supplier No. TRCI780300100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40150-92-3
Molecular Formula C12 H18 O
Molecular Weight (g/mol) 178.27
Synonym 1-(4-Isobutylphenyl)ethanol
IUPAC Name 1-[4-(2-methylpropyl)phenyl]ethanol
SMILES CC(C)Cc1ccc(cc1)C(C)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H18O/c1-9(2)8-11-4-6-12(7-5-11)10(3)13/h4-7,9-10,13H,8H2,1-3H3
Formula Weight 178.1358
Chemical Name or Material 1-(4-Isobutylphenyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.