missing translation for 'onlineSavingsMsg'
Learn More

Alpha-(4-Piperidyl)benzhydrol, TRC

Product Code. 30418201
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30418201 1 g
30414012 50 mg
30416835 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30418201 Supplier Toronto Research Chemicals Supplier No. TRCP4819951G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-46-8
Molecular Formula C18 H21 N O
Molecular Weight (g/mol) 267.37
Synonym Diphenyl(piperidin-4-yl)methanol, Fexofenadine Hydrochloride Imp. E (EP), Terfenadine Imp. I (EP), Azacyclonol, Pipradrol 4-Isomer
IUPAC Name diphenyl(piperidin-4-yl)methanol
SMILES OC(C1CCNCC1)(c2ccccc2)c3ccccc3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H21NO/c20-18(15-7-3-1-4-8-15,16-9-5-2-6-10-16)17-11-13-19-14-12-17/h1-10,17,19-20H,11-14H2
Formula Weight 267.1623
Chemical Name or Material Azacyclonol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.