missing translation for 'onlineSavingsMsg'
Learn More

Alpha,Alpha,3,5-Tetramethyl-benzeneacetonitrile, TRC

Product Code. 30412532
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412532 1 g 1g
30423960 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30412532 Supplier Toronto Research Chemicals Supplier No. TRCT3009001G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93748-07-3
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.25
Synonym 2-(3,5-Dimethylphenyl)-2-methylpropionitrile
IUPAC Name 2-(3,5-dimethylphenyl)-2-methylpropanenitrile
SMILES Cc1cc(C)cc(c1)C(C)(C)C#N

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H15N/c1-9-5-10(2)7-11(6-9)12(3,4)8-13/h5-7H,1-4H3
Formula Weight 173.1204
Chemical Name or Material alpha,alpha,3,5-Tetramethyl-benzeneacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.