missing translation for 'onlineSavingsMsg'
Learn More

Alpha,Alpha,Alpha’,Alpha’-Tetramethyl-5-(dibromomethyl)-1,3-benzenediacetonitrile, TRC

Product Code. 30425443
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30425443 10 mg
30409540 50 mg
30412572 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30425443 Supplier Toronto Research Chemicals Supplier No. TRCT29149510MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1027160-12-8
Molecular Formula C15 H16 Br2 N2
Molecular Weight (g/mol) 384.11
Synonym 1,3-Benzenediacetonitrile, 5-(dibromomethyl)-α1,α1,α3,α3-tetramethyl-, 5-(Dibromomethyl)-α1,α1,α3,α3-tetramethyl-1,3-benzenediacetonitrile
IUPAC Name 2-[3-(2-cyanopropan-2-yl)-5-(dibromomethyl)phenyl]-2-methylpropanenitrile
SMILES CC(C)(C#N)c1cc(cc(c1)C(C)(C)C#N)C(Br)Br

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H16Br2N2/c1-14(2,8-18)11-5-10(13(16)17)6-12(7-11)15(3,4)9-19/h5-7,13H,1-4H3
Formula Weight 381.968
Chemical Name or Material alpha,alpha,alpha',alpha'-Tetramethyl-5-(dibromomethyl)-1,3-benzenediacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.