missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha-Trifluorotoluene, 99+%

Product Code. 10711614 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
500 mL
2.5 L
Packaging:
Plastic bottle
Unit Size:
2.5 L
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10711614 2.5 L Plastic bottle 2.5 L
10723901 50 mL Plastic bottle 50mL
10583301 500 mL Plastic bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10711614 Supplier Thermo Scientific Acros Supplier No. 139770025
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98-08-8 | C7H5F3 | 146.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-08-8
Molecular Formula C7H5F3
Molecular Weight (g/mol) 146.11
MDL Number MFCD00000372
InChI Key GETTZEONDQJALK-UHFFFAOYSA-N
Synonym benzotrifluoride, trifluoromethyl benzene, alpha,alpha,alpha-trifluorotoluene, phenylfluoroform, benzene, trifluoromethyl, benzenyl fluoride, benzylidyne fluoride, usaf ma-16, a,a,a-trifluorotoluene, trifluoro toluene
PubChem CID 7368
ChEBI CHEBI:36810
IUPAC Name trifluoromethylbenzene
SMILES C1=CC=C(C=C1)C(F)(F)F

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -29°C
Color Colorless
Density 1.1900g/mL
Boiling Point 102°C
Flash Point 12°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5CF3
Refractive Index 1.4130 to 1.415
Quantity 2.5 L
Beilstein 05, 290
Merck Index 15, 1113
Specific Gravity 1.19
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in many organic solvents
Formula Weight 146.11
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material α, α, α-Trifluorotoluene, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • a,a,a-Trifluorotoluene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: a,a,a-Trifluorotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.