missing translation for 'onlineSavingsMsg'
Learn More

Alpha,Alpha-Bis[4-(dimethylamino)phenyl]-4-(phenylamino)-1-naphthalenemethanol (Technical Grade), TRC

Product Code. 30010470
Change view
Click to view available options
Quantity:
100 mg
1 g
25 g
Unit Size:
100mg
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010470 25 g 25g
30005889 100 mg 100mg
30007193 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010470 Supplier Toronto Research Chemicals Supplier No. TRCB41641025G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6786-83-0
Molecular Formula C33 H33 N3 O
Molecular Weight (g/mol) 487.63
Synonym C.I. Solvent Blue 4, Aizen Victoria Blue B Base, Baso Blue 645, Brilliant Oil Blue B Base, C.I. 44045B, Fast Oil Blue B Base, Solvent Blue 4, Victoria Blue B Base, Victoria Blue BA Base, Victoria Blue BDP Base, Victoria Blue Base, Victoria Blue Base B, Victoria Blue Base FB, Waxoline Victoria Blue B
IUPAC Name (4-anilino-1-naphthyl)-bis[4-(dimethylamino)phenyl]methanol
SMILES CN(C)c1ccc(cc1)C(O)(c2ccc(cc2)N(C)C)c3ccc(Nc4ccccc4)c5ccccc35

Specifications

Quantity 25 g
InChI Formula InChI=1S/C33H33N3O/c1-35(2)27-18-14-24(15-19-27)33(37,25-16-20-28(21-17-25)36(3)4)31-22-23-32(30-13-9-8-12-29(30)31)34-26-10-6-5-7-11-26/h5-23,34,37H,1-4H3
Formula Weight 487.2624
Chemical Name or Material alpha,alpha-Bis[4-(dimethylamino)phenyl]-4-(phenylamino)-1-naphthalenemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.