missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha'-Dichloro-p-xylene, 98%

Product Code. 10406790 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10406790 100 g Glass bottle 100g
10209323 25 g Glass bottle 25g
10327030 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10406790 Supplier Thermo Scientific Acros Supplier No. 147661000
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 623-25-6 | C8H8Cl2 | 175.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-25-6
MDL Number MFCD00000920
InChI Key ZZHIDJWUJRKHGX-UHFFFAOYSA-N
Synonym 1,4-bis chloromethyl benzene, alpha,alpha'-dichloro-p-xylene, p-xylylene dichloride, p-xylylene chloride, benzene, 1,4-bis chloromethyl, p-bis chloromethyl benzene, benzene, p-bis chloromethyl, a,a'-dichloro-p-xylene, xylylene dichloride, ccris 1772
PubChem CID 12171
IUPAC Name 1,4-bis(chloromethyl)benzene
SMILES C1=CC(=CC=C1CCl)CCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 98.0°C to 102.0°C
Color White to Yellow
Boiling Point 254.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H8Cl2
Linear Formula C6H4(CH2Cl)2
Quantity 100 g
Beilstein 05, 384
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in acetone and methylenechloride,50mg/mL methanol
Molecular Weight (g/mol) 175.06
Formula Weight 175.06
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material α, α'-Dichloro-p-xylene, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • a,a'-Dichloro-p-xylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.