missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha-Dichlorodiphenylmethane, 97%

Product Code. 15499169
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499169 25 g 25g
15489169 5 g 5g
15409179 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15499169 Supplier Thermo Scientific Alfa Aesar Supplier No. L20445.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2051-90-3 | C13H10Cl2 | 237.123 g/mol

α,α-Dichlorodiphenylmethane is a degradation product of of polychloroaromatic insecticides such as Dicofol (D436700) and DDT. It is also used as a catalyst for photodegradation of plastics. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α,α-Dichlorodiphenylmethane is a degradation product of of polychloroaromatic insecticides such as Dicofol (D436700) and DDT. It is also used as a catalyst for photodegradation of plastics. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Soluble in water (reacts), ether, benzene, ethanol, and toluene.

Notes
Stable under recommended storage conditions. Incompatible with strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2051-90-3
Molecular Formula C13H10Cl2
Molecular Weight (g/mol) 237.123
MDL Number MFCD00000811
InChI Key OPTDDWCXQQYKGU-UHFFFAOYSA-N
Synonym dichlorodiphenylmethane, diphenyldichloromethane, dpm halocarbon, benzophenone dichloride, alpha,alpha-dichlorodiphenylmethane, benzene, 1,1'-dichloromethylene bis, methane, dichlorodiphenyl, dichlorodiphenyl methane, dichloro phenyl methyl benzene, unii-81fez29ak6
PubChem CID 16327
IUPAC Name [dichloro(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(Cl)Cl

Specifications

Density 1.242
Boiling Point 180°C to 181°C (17 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.605
Quantity 25 g
UN Number UN3265
Solubility Information Soluble in water (reacts),ether,benzene,ethanol,and toluene.
Formula Weight 237.13
Percent Purity 97%
Chemical Name or Material alpha,alpha-Dichlorodiphenylmethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.