missing translation for 'onlineSavingsMsg'
Learn More

Alpha,Alpha-Dichlorodiphenylmethane, TRC

Product Code. 30382827
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30382827 100 g -
30373890 25 g 25g
30372202 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382827 Supplier Toronto Research Chemicals Supplier No. TRCD434260100G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2051-90-3
Molecular Formula C13H10Cl2
Molecular Weight (g/mol) 237.12
Synonym 1,1'-(Dichloromethylene)bis[benzene], Benzophenone Dichloride, DPM, DPM (halocarbon), Dichlorodiphenylmethane, Diphenyldichloromethane, NSC 37425
SMILES ClC(Cl)(c1ccccc1)c2ccccc2

Specifications

Quantity 100 g
InChI Formula InChI=1S/C13H10Cl2/c14-13(15,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H
Formula Weight 236.016
Chemical Name or Material alpha,alpha-Dichlorodiphenylmethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.