missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha-Dimethylbenzylamine, 96%

Product Code. 11386999
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386999 1 g 1g
11396999 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386999 Supplier Thermo Scientific Alfa Aesar Supplier No. H26845.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 585-32-0 | C9H13N | 135.21 g/mol

Agrochemical Intermediates, dyestuff Intermediate, pharmaceutical Intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agrochemical Intermediates, dyestuff Intermediate, pharmaceutical Intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-32-0
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00134680
InChI Key KDFDOINBXBEOLZ-UHFFFAOYSA-N
Synonym cumylamine, alpha,alpha-dimethylbenzylamine, 1-methyl-1-phenyl-ethylamine, 1-methyl-1-phenylethylamine, 2-phenyl-2-propanamine, alpha-aminoisopropylbenzene, benzenemethanamine, alpha,alpha-dimethyl, dimethyl-aminophenylmethane, cumyl amine, phenisopropyl amine
PubChem CID 68509
IUPAC Name 2-phenylpropan-2-amine
SMILES CC(C)(N)C1=CC=CC=C1

Specifications

Density 0.94
Boiling Point 197°C
Flash Point 72°C (162°F)
Odor Amine-like
Refractive Index 1.52
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 135.21
Percent Purity 96%
Chemical Name or Material alpha,alpha-Dimethylbenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.