missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha-Dimethylphenylacetic acid

Product Code. 11393018
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393018 2 g 2g
11383018 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393018 Supplier Thermo Scientific Alfa Aesar Supplier No. 032884.04
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 826-55-1 | C10H12O2 | 164.20 g/mol

α,α-Dimethylphenylacetic acid is used in the synthesis of active pharmaceutical ingredient, fexofenadine hydrochloride, which is used as antihistamine. Further, it acts as pharmaceutical intermediates and as chemical intermediates in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 826-55-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00014332
InChI Key YYEROYLAYAVZNW-UHFFFAOYSA-N
Synonym 2-methyl-2-phenylpropionic acid, 2-phenylisobutyric acid, alpha,alpha-dimethylphenylacetic acid, unii-dr8huh2gyk, dimethylphenylacetic acid, dr8huh2gyk, 2-methyl-2-phenyl-propionic acid, hydratropic acid, .alpha.-methyl, benzeneacetic acid,.alpha.-dimethyl, propionic acid, 2-methyl-2-phenyl
PubChem CID 13222
IUPAC Name 2-methyl-2-phenylpropanoic acid
SMILES CC(C)(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 80°C to 82°C
Boiling Point 146.5°C
Quantity 2 g
Solubility Information Insoluble in water.
Formula Weight 164.21
Physical Form Crystalline
Chemical Name or Material alpha,alpha-Dimethylphenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.