missing translation for 'onlineSavingsMsg'
Learn More

alpha-Asarone, MedChemExpress

Product Code. 16442549
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442549 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16442549 Supplier Medchem Express Supplier No. HYN0700
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress alpha-Asarone (α-Asarone) is one of the main psychoactive compounds, and possesses an antidepressant-like activity in mice.

alpha-Asarone (α-Asarone) is one of the main psychoactive compounds, and possesses an antidepressant-like activity in mice.

Specifications

Chemical Name or Material alpha-Asarone
CAS 2883-98-9
Color White
Purity Grade Notes Research
Molecular Formula C12H16O3
Quantity 1 g
Synonym α-Asarone trans-Asarone
Solubility Information DMSO : 100 mg/mL (480.19 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (3.22 mM; Need ultrasonic)
SMILES C/C=C/C1=C(OC)C=C(OC)C(OC)=C1
Molecular Weight (g/mol) 208.25
Formula Weight 208.25
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.