missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-2,3,4,5,6-pentafluorotoluene, TRC

Product Code. 30136128
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136128 5 g 5g
17961685 25 g 25g
17993075 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136128 Supplier Toronto Research Chemicals Supplier No. TRCB6870405G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1765-40-8
Molecular Formula C7 H2 Br F5
Molecular Weight (g/mol) 260.99
Synonym 1-(Bromomethyl)-2,3,4,5,6-pentafluorobenzene (ACI), Benzene, (bromomethyl)pentafluoro- (9CI), Toluene, α-bromo-2,3,4,5,6-pentafluoro- (6CI, 7CI, 8CI), (Bromomethyl)pentafluorobenzene, (Pentafluorophenyl)methyl bromide, 1-(Bromomethyl)pentafluorobenzene, 2,3,4,5,6-Pentafluoro-α-bromotoluene, 2,3,4,5,6-Pentafluorobenzyl bromide, 2,3,4,5,6-Pentafluorophenylmethyl bromide, NSC 96888, Pentafluorobenzyl bromide, α-Bromo-2,3,4,5,6-pentafluorotoluene, α-Bromo-pentafluorotoluene
IUPAC Name 1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene
SMILES Fc1c(F)c(F)c(CBr)c(F)c1F

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H2BrF5/c8-1-2-3(9)5(11)7(13)6(12)4(2)10/h1H2
Formula Weight 259.926
Chemical Name or Material a-Bromo-2,3,4,5,6-pentafluorotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.